Learn More
ARMC6 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25558-1-AP-20UL
 View more versions of this product
Product Code. 18039475
  • € 145.00 / 20µL
Description
The ARMC6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARMC6. This antibody recognizes human antigen. The ARMC6 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ARMC6
500 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
armadillo repeat containing 6, ARMC6, R30923_1
20 μL
93436
Store at -20°C. Stable for one year after shipment.