Learn More
ARMC6 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25558-1-AP-150UL
This item is not returnable.
View return policy
Description
The ARMC6 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARMC6. This antibody recognizes human antigen. The ARMC6 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ARMC6 | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| armadillo repeat containing 6, ARMC6, R30923_1 | |
| 150 μL | |
| 93436 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |