missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARMCX4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA551388

Product Code. 15799340

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GRAQSVRTLA RNGSTVKMET VVGVQSQTLA INEAEIKTKP QVEIGAETGA RSGPRAEVET KATAIAIHRA NSQAKAMVGA EPETQSESKV VAGTLVMTEA VTLTEVKAKA REVAMKEAVT QTDAEAGKIV KKEAVTQTKA Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 53%.

The product of this gene belongs to the armadillo repeat-containing family of proteins, which interact with other proteins in a variety of cellular processes. The function of this family member is currently unknown. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq, Jan 2012].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ARMCX4
Polyclonal
Unconjugated
ARMCX4
armadillo repeat containing, X-linked 4; armadillo repeat containing, X-linked 4 pseudogene; armadillo repeat-containing X-linked protein 4; ARMCX4; CXorf35; GASP4
Rabbit
Antigen Affinity Chromatography
RUO
100131755
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5H9R4
ARMCX4
Recombinant protein corresponding to Human ARMCX4. Recombinant protein control fragment (Product #RP-92015).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.