missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARMX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536203
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 66 kDa. Purity is >95% by SDS-PAGE.
The armadillo (ARM) repeat family of proteins are related to the Drosophila melanogaster armadillo protein, a protein essential for wingless signal transduction. ARM proteins are involved in a variety of processes such as cell migration, cell proliferation, tissue maintenance and tumorigenesis, and they also function in signal transduction and the maintenance of overall cell structure. ARMCX2 (armadillo repeat containing, X-linked 2), also known as ALEX2, is a 632 amino acid single-pass membrane protein that contains three ARM repeats and is highly expressed in testis, brain, ovary, heart, colon, spleen and prostate, where it is thought to play a role in tumor suppression. The gene encoding ARMCX2 maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1, 000 genes.
Specifications
| ARMX2 | |
| Polyclonal | |
| Unconjugated | |
| ARMCX2 | |
| 3230401N03Rik; AI043003; ALEX2; ALEX2 protein; ARM protein lost in epithelial cancers on chromosome X 2; armadillo repeat containing, X-linked 2; armadillo repeat protein ALEX2; armadillo repeat-containing X-linked protein 2; ARMCX2; GASP9; Kiaa0512; Protein ALEX2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 367903, 67416, 9823 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6A058, Q7L311 | |
| ARMCX2 | |
| Synthetic peptide corresponding to amino acids 312-368 of human ARMCX2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction