missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ARN 11391
GREENER_CHOICE

Product Code. 18735825
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18735825

Brand: Tocris Bioscience™ 7909/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potentiator of the IP3-gated calcium channel ITPR1

ARN 11391 is a potentiator of the IP3-gated calcium channel ITPR1 (Inositol 1,4,5-trisphosphate (IP3) receptor type 1). It amplifies intracellular calcium ion signaling by acting directly on ITPR1. ARN 11391 also potentiates calcium signaling in cells expressing mutant ITPR1 which is associated with SCA29, a form of spinocerebellar ataxia.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Ethyl-4,5,6,7-tetrahydro-N-[2-[(tetrahydro-2 H-pyran-2-yl)methoxy]phenyl]-2 H-indazole-3-carboxamide
CAS 1214569-31-9
Quantity 10 mg
Target IP3R1
Molecular Formula C22H29N3O3
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.