missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARNT/HIF-1 beta Antibody (H1beta234), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-124AF594
This item is not returnable.
View return policy
Description
ARNT/HIF-1 beta Monoclonal specifically detects ARNT/HIF-1 beta in Human, Mouse, Rat, Bovine, Ferret, Primate, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.
Specifications
| ARNT/HIF-1 beta | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ARNT | |
| Fusion protein containing amino acids 496-789 of human HIF-1 beta. [Uniprot: P27540] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Ferret, Primate, Sheep | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| H1beta234 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting | |
| ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, HIF-1 Beta, HIF1B, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, nuclear translocator | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Autophagy, Cancer, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Transcription Factors and Regulators | |
| 405.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction