missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aromatase Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1596B
This item is not returnable.
View return policy
Description
Aromatase Polyclonal specifically detects Aromatase in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Aromatase | |
| Polyclonal | |
| Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| P11511 | |
| CYP19A1 | |
| A synthetic peptide made to a C-terminal portion of the human Aromatase protein (between residues 400-502). [UniProt# P11511] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human and Primate. Immunogen has 91% homology to horse, 90% homology to cow and rabbit, 83% homology to pig, and 81% homology to sheep, goat, and dog proteins. | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ARO1EC 1.14.14.1, Aromatase, AROsubfamily XIX (aromatization of androgens), CYARMGC104309, CYPXIX, cytochrome P450, family 19, subfamily A, polypeptide 1, Cytochrome P-450AROM, Estrogen synthase, microsomal monooxygenase | |
| Rabbit | |
| 55 kDa | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Lipid and Metabolism, Neuroscience | |
| 1588 | |
| Human, Mouse, Rat, Primate, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction