Learn More
Aromatase Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99064MFV450
Product Code. 18550213
  • € 428.00 / 0.10mL
Description
Aromatase Polyclonal antibody specifically detects Aromatase in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Aromatase.
0.1 mL
Primary
Antigen and protein A Affinity-purified