missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: CRDHLRVNLK FLTRTRSTVA ESVSQQILRQ NRGRKACDRP VAKTKKKKAE GTVFTEEDFQ KFQQEYFGS Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.
AROS, a nucleolar protein with an unknown function, interacts with extra ribosomal protein RPS19, playing a role in the signaling pathways that regulate rRNA transcription. The deduced amino acid sequence of AROS, derived from cDNA, defines an isoelectric point of 11.3. No known functional motifs were found in AROS except a short polylysine tract embedded in a putative nucleolar localization signal. At a basal level the protein is expressed ubiquitously with a significantly high level of expression in some tissues. Human AROS gene is mapped to chromosome 22q13.1.
Specifications
Specifications
| Antigen | AROS |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RPS19BP1 |
| Gene Accession No. | Q86WX3 |
| Gene Alias | 2510038A11Rik; 40S ribosomal protein S19-binding protein 1; Active regulator of SIRT1; AI604923; AROS; dJ1104E15.4; homolog of mouse S19 binding protein; RGD1562823; ribosomal protein S19 binding protein 1; RP5-1104E15.4; RPS19-binding protein 1; RPS19BP1; S19 binding protein; S19BP |
| Gene Symbols | RPS19BP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
