missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15471
This item is not returnable.
View return policy
Description
ARPC1A Polyclonal specifically detects ARPC1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ARPC1A | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
| actin binding protein (Schizosaccharomyces pombe sop2-like), actin related protein 2/3 complex, subunit 1A (41 kD), actin related protein 2/3 complex, subunit 1A, 41kDa, actin-related protein 2/3 complex subunit 1A, SOP2Hs, SOP2LArc40, SOP2-like protein | |
| Rabbit | |
| 42 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Xenopus laevis (92%) and Xenopus tropicalis (92%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q92747 | |
| ARPC1A | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human ARPC1A. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 10552 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction