missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ARPC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29648
This item is not returnable.
View return policy
Description
ARPC2 Polyclonal antibody specifically detects ARPC2 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ARPC2 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| actin related protein 2/3 complex, subunit 2 (34 kD), actin related protein 2/3 complex, subunit 2, 34kDa, ARC34actin-related protein 2/3 complex subunit 2, Arp2/3 complex 34 kDa subunit, ARP2/3 protein complex subunit 34, p34-ARC, PNAS-139 | |
| Between 175 and 225 | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 10109 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction