missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARPC5 Antibody (OTI2G1), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71550B
This item is not returnable.
View return policy
Description
ARPC5 Monoclonal specifically detects ARPC5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ARPC5 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| ARPC5 | |
| Full length recombinant protein of human ARPC5 (NP_005708) produced in HEK293T cells. | |
| 0.1 mL | |
| 10092.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI2G1 | |
| Western Blot, Immunohistochemistry | |
| actin related protein 2/3 complex, subunit 5, 16kDa, ARC16subunit 5 (16 kD), MGC88523 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction