Learn More
ARPC5 Mouse anti-Human, Clone: OTI2G1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505696
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes one of seven subunits of the human Arp2/3 protein complex. The Arp2/3 protein complex has been implicated in the control of actin polymerization in cells and has been conserved through evolution. The exact role of the protein encoded by this gene, the p16 subunit, has yet to be determined.Specifications
| ARPC5 | |
| Monoclonal | |
| Unconjugated | |
| ARPC5 | |
| ARC16, dJ127C7.3, p16-Arc | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10092 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O15511 | |
| ARPC5 | |
| Full length human recombit protein of human ARPC5 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.