missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549513
This item is not returnable.
View return policy
Description
ARR3 Polyclonal Antibody for Western Blot
May play a role in an as yet undefined retina-specific signal transduction. Could binds to photoactivated-phosphorylated red/green opsins.
Specifications
| ARR3 | |
| Polyclonal | |
| Unconjugated | |
| Arr3 | |
| ARR3; Arr4; arrestin 3; arrestin 3 retinal (X-arrestin); arrestin 3, retinal; arrestin 3, retinal (X-arrestin); arrestin 4; Arrestin C; ArrestinC; arrestin-C; ARRX; Car; Carfl; cArr; C-arrestin; Cone arrestin; retinal cone arrestin-3; X-arrestin | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 170735, 407 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P36575, Q9EQP6 | |
| Arr3 | |
| KLH conjugated synthetic peptide between 77-109 amino acids from the human region of human ARR3. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction