missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARRB2, Goat anti-Human, Polyclonal Antibody, Abnova™
Goat Polyclonal Antibody
Brand: Abnova PAB6785.100ug
This item is not returnable.
View return policy
Description
Sequence: C-HDHIPLPRPQSSpecifications
| ARRB2 | |
| Polyclonal | |
| Unconjugated | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| ARB2/ARR2/BARR2/DKFZp686L0365 | |
| Goat | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms NP_004304.1 and NP_945355.1. No crossreactivity is expected with Arrestin beta 1. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.5 mg/mL | |
| ELISA (1:32000) Western Blot (0.3-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-6 ug/mL) The optimal working dilution should be determined by the end user. | |
| arrestin, beta 2 | |
| ARRB2 | |
| A synthetic peptide corresponding to internal region of human ARRB2. | |
| RUO | |
| 409 | |
| Human, Mouse, Rat | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction