Learn More
ARRDC1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21030-1-AP-150UL
 View more versions of this product
Product Code. 18020336
  • € 410.00 / 150µL
Description
The ARRDC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARRDC1. This antibody recognizes human antigen. The ARRDC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
ARRDC1
350 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
ARRDC1, arrestin domain containing 1
150 μL
92714
Store at -20°C. Stable for one year after shipment.