Learn More
Abeba™ Arrow 8210 Shoes
Product Code. 15693058
Click to view available options
Size:
36
37
38
39
41
42
44
45
46
47
Unit Size:
1 pair
Product Code. 15693058
Brand: Abeba™ 8210/44
Only null left
Add to basket
Add to basket
Antistatic with heel patch microfiber. Sanitized™ treated.
  • CE EN ISO 20347:2012 OB A, E, FO SRA
  • Smooth leather
  • Black
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Microfiber heel patch
  • Lining microfiber, respiration absorbing
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, FO SRA
Color Black
Liner Material Microfiber
Material Leather
Size 44
Sole Features Slip-resistant
Sole Material Polyurethane
Style 8210