Learn More
ARS-1323, MedChemExpress
Product Code. 16422628
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. 16422628
Brand: Medchem Express HYU00416
MedChemExpress ARS-1323, the racemate of ARS-1620, is a novel inhibitor of mutant K-ras G12C extracted from patent WO 2015054572 A1.
ARS-1323, the racemate of ARS-1620, is a novel inhibitor of mutant K-ras G12C extracted from patent WO 2015054572 A1.
Specifications
Chemical Name or Material ARS-1323
CAS 1698024-73-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H17ClF2N4O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (232.10 mM; Need ultrasonic)
SMILES O=C(C=C)N1CCN(C2=NC=NC3=C2C=C(Cl)C(C4=C(O)C=CC=C4F)=C3F)CC1
Molecular Weight (g/mol) 430.84
Formula Weight 430.84
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only