missing translation for 'onlineSavingsMsg'
Learn More

Arsenic Standard: 1000 μg/g As in 75 cSt Hydrocarbon Oil, VHG™

Product Code. 17900936
Click to view available options
Quantity:
50 g
200 g
400 g
Unit Size:
200g
400g
50g
This item is not returnable. View return policy

Product Code. 17900936

Brand: VHG Labs VHGOAS1000200G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Arsenic Standard: 1000 μg/g As in 75 cSt Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arsenic
CAS 7440-38-2
Methods ICP/XRF/RDE
Analyte or Component Names Arsenic
Shipping Condition Room Temperature
Molecular Formula As
Certifications/Compliance ISO 17034
Synonym Arsenicals, Arsenic, inorganic, Arsen
InChI Formula InChI=1S/As
SMILES [As]
IUPAC Name arsenic
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 74.92
Quantity 200 g
Formula Weight 74.922 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.