missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARSH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103618
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARSH.
Sulfatases, such as ARSH, hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans, and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling, and degradation of macromolecules.
Specifications
| ARSH | |
| Polyclonal | |
| Unconjugated | |
| ARSH | |
| ARSH; arylsulfatase family member H; arylsulfatase family, member H; arylsulfatase H; arylsulfatase H; arylsulfatase family member H; ASH; sulfatase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 347527 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5FYA8 | |
| ARSH | |
| A synthesized peptide derived from human ARSH(Accession Q5FYA8), corresponding to amino acid residues R487-P537. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction