Learn More
Thermo Scientific™ ART™ Non-Filtered Low Retention Universal Pipette Tips
Specifically designed for eliminating sample retention within the pipette tip
Brand: Thermo Scientific™ 3581-05
Additional Details : Weight : 1.61100kg
Description
- Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
- Crystal clear tips allow you to easily see transparent samples
- Reference marks on many Thermo Scientific ART tips provide a quick and convenient guide to visually establish whether you are pipetting desired volume of liquid
- All Pure tips are racked, presterilized and certified RNase-, DNase-, DNA- and pyrogen-free
- Other Thermo Scientific racked and bulk tips are nonsterile and autoclavable
Specifications
100 to 1000 μL | |
Sterile | |
Autoclavable | |
Rack | |
ART |
Non-Filtered | |
Clear | |
SoftFit | |
Low Retention |