missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART1 Antibody (OTI6E2), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70211FR
This item is not returnable.
View return policy
Description
ART1 Monoclonal specifically detects ART1 in Human samples. It is validated for Western Blot.
Specifications
| ART1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| ART1 | |
| Recombinant protein fragment corresponding to amino acids 83-295 of human ART1 (NP_004305) produced in E.coli. | |
| 0.1 mL | |
| 417.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI6E2 | |
| Western Blot | |
| ADP-ribosyltransferase 1, ADP-ribosyltransferase 2, CD296, GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1, MGC133217, mono(ADP-ribosyl)transferase 1, RT6 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction