missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART2.2/Art2b Antibody (Nika102), DyLight 755, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-65853IR
This item is not returnable.
View return policy
Description
ART2.2/Art2b Monoclonal specifically detects ART2.2/Art2b in Mouse samples. It is validated for Flow Cytometry.
Specifications
| ART2.2/Art2b | |
| Monoclonal | |
| DyLight 755 | |
| ADP-ribosyltransferase 2b, Rt6, Rt-6, Rt6.2, Rt6-2 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 11872 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| Nika102 | |
| Flow Cytometry | |
| ART2B | |
| pME.CD8LF-ART2.2 | |
| 0.125 mL | |
| Primary | |
| NB100-65853 recognizes the gene product of the mouse ADP-ribosyltransferase (ART) 2.2 gene, an ortholog of rat RT6. ART2.2 expressed exclusively by mature T-cells. Levels of ART2.2 expression vary between inbred strains of mice and is absent in NZW mice, in which the ART2.2 gene is deleted (PMID: 10570289, Koch-Nolte et al. 1999). | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction