missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00143JF646
This item is not returnable.
View return policy
Description
ART3 Polyclonal antibody specifically detects ART3 in Human samples. It is validated for ELISA
Specifications
| ART3 | |
| Polyclonal | |
| ELISA | |
| ADP-ribosyltransferase 3, EC 2.4.2.31, ecto-ADP-ribosyltransferase 3, FLJ26404, Mono(ADP-ribosyl)transferase 3, mono-ADP-ribosyltransferase, NAD(P)(+)--arginine ADP-ribosyltransferase 3, TMART | |
| Produced in rabbits immunized with purified, recombinant Human ART3 (Accession#: AAH08397.1; Met1-Ser362). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 419 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction