missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ART4, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00000420-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MGPLINRCKKILLPTTVPPATMRIWLLGGLLPFLLLLSGLQRPTEGSEVAIKIDFDFAPGSFDDQYQGCSKQVMEKLTQGDYFTKDIEAQKNYFRMWQKAHLAWLNQGKVLPQNMTTTHAVAILFYTLNSNVHSDFTRAMASVARTPQQYERSFHFKYLHYYLTSAIQLLRKDSIMENGTLCYEVHYRTKDVHFNAYTGATIRFGQFLSTSLLKEEAQEFGNQTLFTIFTCLGAPVQYFSLKKEVLIPPYELFKVINMSYHPRGDWLQLRSTGNLSTYNCQLLKASSKKCIPDPIAIASLSFLTSVIIFSKSRVSpecifications
| ART4 | |
| Polyclonal | |
| No additive | |
| NM_021071.2 | |
| ART4 | |
| ART4 (NP_066549.2, 1 a.a. ∼ 314 a.a) full-length human protein. | |
| RUO | |
| 420 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation | |
| Unconjugated | |
| ADP-ribosyltransferase 4 (Dombrock blood group) | |
| CD297/DO/DOK1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur