missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemin Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-23618MFV500
This item is not returnable.
View return policy
Description
Artemin Polyclonal antibody specifically detects Artemin in Human samples. It is validated for Western Blot
Specifications
| Artemin | |
| Polyclonal | |
| Western Blot | |
| artemin, ENOVIN, EVNneurotrophic factor, NBN, Neublastin | |
| Reacts with the N terminus of Artemin. | |
| 0.1 mL | |
| Cancer, Growth and Development, Neuronal Cell Markers, Neuroscience | |
| 9048 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction