missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Artemin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142605
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize all reported isoforms (NP_003967.2; NP_476501.1; NP_476431.2). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
The protein encoded by this gene is a member of the glial cell line-derived neurotophic factor (GDNF) family of ligands which are a group of ligands within the TGF-beta superfamily of signaling molecules. GDNFs are unique in having neurotrophic properties and have potential use for gene therapy in neurodegenerative disease. Artemin has been shown in culture to support the survival of a number of peripheral neuron populations and at least one population of dopaminergic CNS neurons. Its role in the PNS and CNS is further substantiated by its expression pattern in the proximity of these neurons. This protein is a ligand for the RET receptor and uses GFR-alpha 3 as a coreceptor. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| Artemin | |
| Polyclonal | |
| Unconjugated | |
| ARTN | |
| Artemin; ARTN; Enovin; EVN; NBN; neublastin; neurotrophic factor | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 9048 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q5T4W7 | |
| ARTN | |
| Peptide with sequence C-NSTWRTVDRLS. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction