missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemis Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-183SS
This item is not returnable.
View return policy
Description
Artemis Polyclonal specifically detects Artemis in Human samples. It is validated for Western Blot.
Specifications
| Artemis | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| DCLRE1C | |
| A synthetic peptide made to residues 677-692 [GESIAVKKRKCSLLDT] of the human Artemis protein. | |
| Affinity Purified | |
| Cancer, DNA Repair, Non homologous end joining | |
| 64421 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:250-1:500 | |
| ASCID, DCLREC1C, DNA cross-link repair 1C, DNA cross-link repair 1C protein, EC 3.1, FLJ36438, hSNM1C, protein artemis, Protein A-SCID, PSO2 homolog, RS-SCID, severe combined immunodeficiency, type a (Athabascan), SNM1 homolog C, SNM1CS. cerevisiae), SNM1-like protein | |
| Rabbit | |
| 66 kDa | |
| 0.025 mg | |
| Primary | |
| Artemis | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction