missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemis Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44276IR
This item is not returnable.
View return policy
Description
Artemis Polyclonal antibody specifically detects Artemis in Mouse, Rat samples. It is validated for Western Blot
Specifications
| Artemis | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, DNA Repair, Non homologous end joining | |
| Mouse, Rat |
| Western Blot | |
| DyLight 755 | |
| Immunogen affinity purified | |
| 64421 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction