missing translation for 'onlineSavingsMsg'
Learn More

Artemis Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18780500 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18780500 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18780500 Supplier Novus Biologicals Supplier No. NB100183MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Artemis Polyclonal antibody specifically detects Artemis in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Artemis
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ASCID, DCLREC1C, DNA cross-link repair 1C, DNA cross-link repair 1C protein, EC 3.1, FLJ36438, hSNM1C, protein artemis, Protein A-SCID, PSO2 homolog, RS-SCID, severe combined immunodeficiency, type a (Athabascan), SNM1 homolog C, SNM1CS. cerevisiae), SNM1-like protein
Host Species Rabbit
Immunogen A synthetic peptide made to residues 677-692 [GESIAVKKRKCSLLDT] of the human Artemis protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, DNA Repair, Non homologous end joining
Primary or Secondary Primary
Gene ID (Entrez) 64421
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.