missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Artemis Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111646
This item is not returnable.
View return policy
Description
Immunogen sequence: WDSQSDTVLL SSQERNSGDI TSLDKADYRP TIKENIPASL MEQNVICPKD TYSDLKSRDK DVTIVPSTGE PTTLS Highest antigen sequence indentity to the following orthologs: Rat - 57%, Mouse - 57%.
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
Specifications
| Artemis | |
| Polyclonal | |
| Unconjugated | |
| DCLRE1C | |
| 9930121L06Rik; AI661365; Art; ARTEMIS; ASCID; A-SCID; Dclre1c; DCLRE1C protein; DCLREC1C; DNA cross-link repair 1A, PSO2 homolog; DNA cross-link repair 1C; DNA cross-link repair 1C (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1C protein; DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae); FLJ11360; hSNM1C; mArt; Protein artemis; Protein A-SCID; RS SCID; RS-SCID; SCIDA; severe combined immunodeficiency, type a (Athabascan); SNM1 homolog C; SNM1C; SNM1C antibody; Snm1l; SNM1-like protein; tein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64421 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96SD1 | |
| DCLRE1C | |
| Recombinant Protein corresonding to Human Artemis. Recombinant protein control fragment (Product #RP-107685). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction