Learn More
Artemisinin, 98%, Thermo Scientific™
€ 129.00 - € 378.99
Chemical Identifiers
| CAS | 63968-64-9 |
|---|---|
| Molecular Formula | C15H22O5 |
| Molecular Weight (g/mol) | 282.336 |
| MDL Number | MFCD00081057 |
| InChI Key | BLUAFEHZUWYNDE-DKGJTOOQSA-N |
| Synonym | artemisinine, artemisinin, gnf-pf-5341, artemisinin, 18, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.0^ 4,13 .0^ 8,13 hexadecan-10-one, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.04,13.08,13 hexadecan-10-one |
| PubChem CID | 9838675 |
| SMILES | CC1CCC2C(C(=O)OC3C24C1CCC(O3)(OO4)C)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15456889
|
Thermo Scientific Alfa Aesar
J65406.03 |
1g |
€ 132.00 € 129.00 / 1g Save € 3.00 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15466889
|
Thermo Scientific Alfa Aesar
J65406.06 |
5g |
€ 387.00 € 378.99 / 5g Save € 8.01 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble to 100mM in DMSO and to 75mM in ethanol
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents, heat. Store in refrigerator.
Chemical Identifiers
| 63968-64-9 | |
| 282.336 | |
| BLUAFEHZUWYNDE-DKGJTOOQSA-N | |
| 9838675 |
| C15H22O5 | |
| MFCD00081057 | |
| artemisinine, artemisinin, gnf-pf-5341, artemisinin, 18, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.0^ 4,13 .0^ 8,13 hexadecan-10-one, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.04,13.08,13 hexadecan-10-one | |
| CC1CCC2C(C(=O)OC3C24C1CCC(O3)(OO4)C)C |
Specifications
| 63968-64-9 | |
| MFCD00081057 | |
| artemisinine, artemisinin, gnf-pf-5341, artemisinin, 18, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.0^ 4,13 .0^ 8,13 hexadecan-10-one, 1s,4s,5r,8s,9r,12s,13r-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo 10.3.1.04,13.08,13 hexadecan-10-one | |
| BLUAFEHZUWYNDE-DKGJTOOQSA-N | |
| CC1CCC2C(C(=O)OC3C24C1CCC(O3)(OO4)C)C | |
| 9838675 | |
| 99% |
| C15H22O5 | |
| 14,817 | |
| Soluble to 100mM in DMSO and to 75mM in ethanol | |
| +76° (c=0.5 in Methanol) | |
| 282.336 | |
| 282.33 | |
| Artemisinin |
Safety and Handling
missing translation for 'rtecsNumber' : KD4170000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures