missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARTS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111151
This item is not returnable.
View return policy
Description
Immunogen sequence: FQLVSIGKLS IEKALDLSLY LKHETEIMPV FQGLNELIPM YKLMEKRDMN EVETQFKAFL IRLLRDLIDK QTWTDEGSVS ERMLRSQLLL LACVHNYQPC VQRAEGYFRK WKESNGNLS Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 88%.
The endoplasmic reticulum (ER) aminopeptidase 1 (ERAP1) is a 120 kDa protein localized to the lumen of the ER, which removes NH2-terminal residues from many antigenic precursors for MHC class I peptide presentation. Peptides that are presented by MHC class I on the surface of a cell must be 8-11 residues long, and ERAP1 specifically trims peptides of 9 amino acids or more. ERAP1 is also induced by interferon- gamma. The gene encoding human ERAP1 maps to chromosome 5q15. ERAP1 has previously been characterized as adipocyte-derived leucine aminopeptidase (A-LAP), puromycin-insensitive leucine-specific aminopeptidase (PILS-AP) and aminopeptidase regulator of TNFR1 shedding (ARTS-1). A-LAP is thought to inactivate several bioactive peptides, including angiotensin II and, subsequently, may be involved in the regulation of blood pressure. PILS-AP is described as playing a role in angiogenesis by regulating the proliferation and migration of endothelial cells, and ARTS-1 is characterized as a TNFR1 binding protein that promotes TNFR1 shedding. Further research will be necessary to fully elucidate the functions of this protein.
Specifications
| ARTS1 | |
| Polyclonal | |
| Unconjugated | |
| Erap1 | |
| adipocyte-derived leucine aminopeptidase; ALAP; A-LAP; aminopeptidase PILS; aminopeptidase regulator of TNFR1 shedding; Appils; Arts1; ARTS-1; CTD-2260A17.2; endoplasmic reticulum aminopeptidase 1; endoplasmic reticulum aminopeptidase 1 delta-Exon-11 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-13 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-14 isoform; endoplasmic reticulum aminopeptidase 1 delta-Exon-15 isoform; endoplasmic reticulum aminopeptidase associated with antigen processing; ERAAP; ERAAP1; ER-aminopeptidase 1; ERAP1; KIAA0525; leucyl-specific aminopeptidase PILS; PILSA; PILSAP; PILS-AP; Puromycin-insensitive leucyl-specific aminopeptidase; type 1 tumor necrosis factor receptor shedding aminopeptidase regulator; UNQ584/PRO1154; VEGF-induced aminopeptidase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51752 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZ08 | |
| Erap1 | |
| Recombinant Protein corresonding to Human ARTS1. Recombinant protein control fragment (Product #RP-104752). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction