missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arylsulfatase A/ARSA Antibody (001), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89502CL1
This item is not returnable.
View return policy
Description
Arylsulfatase A/ARSA Monoclonal antibody specifically detects Arylsulfatase A/ARSA in Human samples. It is validated for Western Blot
Specifications
| Arylsulfatase A/ARSA | |
| Monoclonal | |
| CoraFluor 1 | |
| arylsulfatase A, ASA, Cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Arylsulfatase A/ARSA (Uniprot#: P15289-1; Met 1-Ala 507). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 410 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Western Blot | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction