missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arylsulfatase A/ARSA Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98210JF646
This item is not returnable.
View return policy
Description
Arylsulfatase A/ARSA Polyclonal antibody specifically detects Arylsulfatase A/ARSA in Mouse samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Arylsulfatase A/ARSA | |
| Polyclonal | |
| Western Blot, ELISA, Immunoprecipitation | |
| arylsulfatase A, ASA, Cerebroside-sulfatase, EC 3.1.6, EC 3.1.6.8, MLD | |
| Produced in rabbits immunized with purified, recombinant Mouse Arylsulfatase A/ARSA (Accession#: NP_033843.2; Met 1-Ser 506) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 410 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunoprecipitation | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction