missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASAH2/N-acylsphingosine Amidohydrolase-2 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76360AF647
This item is not returnable.
View return policy
Description
ASAH2/N-acylsphingosine Amidohydrolase-2 Polyclonal antibody specifically detects ASAH2/N-acylsphingosine Amidohydrolase-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| ASAH2/N-acylsphingosine Amidohydrolase-2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 647 | |
| ASAH2 N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 2, BCDase, HNAC1, LCDase, NCDase, N-CDase | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human ASAH2. The immunogen is located within the last 50 amino acids of ASAH2. Amino Acid Squence: YRIRYFGHNRKQDILK | |
| 0.1 mL | |
| Signal Transduction | |
| 56624 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction