Learn More
ASAM Antibody (001), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90634MFV500
This item is not returnable.
View return policy
Description
ASAM Monoclonal antibody specifically detects ASAM in Mouse samples. It is validated for ELISA