Learn More
ASAM Antibody (001), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90634MFV500
Product Code. 18451839
  • € 629.00 / 0.10mL
Description
ASAM Monoclonal antibody specifically detects ASAM in Mouse samples. It is validated for ELISA