Learn More
Invitrogen™ ASAM Recombinant Rabbit Monoclonal Antibody (001)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA529168
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse ASAM.
May be involved in the cell-cell adhesion. May play a role in adipocyte differentiation and development of obesity. Is required for normal small intestine development.
Specifications
| ASAM | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8R373 | |
| Clmp | |
| Recombinant Mouse ASAM/ACAM protein (Met1-Met232). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| ELISA | |
| 001 | |
| Unconjugated | |
| Clmp | |
| 9030425E11Rik; ACAM; Adipocyte adhesion molecule; adipocyte-specific adhesion molecule; adipocyte-specific protein 5; Asam; ASP5; AW557819; CAR-like membrane protein; Clmp; Coxsackie- and adenovirus receptor-like membrane protein; CSBM; CSBS; CXADR like membrane protein; CXADR-like membrane protein; FLJ22415; LOW QUALITY PROTEIN: CXADR-like membrane protein; Ol16; UNQ318/PRO363; visceral adipose tissue-specific transmembrane protein OL-16 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 71566 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.