missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ASAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552796
This item is not returnable.
View return policy
Description
Immunogen sequence: STPRDKQRLS YGAFTNQIFV STSTDSPTSP TTEAPPLPPR NAGKGNDGGP SSSSKTTNKF EGLSQQSSTS SAKTALGPRV LPKLPQKVAL RKT Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 95%.
ASAP1, also known as Development and differentiation enhancing factor 1, is a protein with two ANK repeats, an Arf-GAP domain, a PH domain and a SH3 domain. ASAP1 is a homodimer known to interact with SRC and CRK and its activity is stimulated by phosphatidylinositol 4,5-biphosphate (PIP2). It possesses phosphatidylinositol 4,5-biphosphate-dependent GTPase-activating protein activity for ARF1 (ADP ribosylation factor 1) and ARF5 and a lesser activity towards ARF6. It may coordinate membrane trafficking with cell growth or actin cytoskeleton remodeling by binding to both SRC and PIP2 and may also function as a signal transduction protein involved in the differentiation of fibroblasts into adipocytes and possibly other cell types. It is widely expressed in most of the tissues. ASAP1 is a protooncogene involved in invasive breast cancer and in other types of invasive and malignant tumors, such as uveal melanomas.
Specifications
| ASAP1 | |
| Polyclonal | |
| Unconjugated | |
| ASAP1 | |
| 130 kDa phosphatidylinositol 4,5-biphosphate-dependent ARF1 GTPase-activating protein; 130 kDa phosphatidylinositol 4,5-bisphosphate-dependent ARF1 GTPase-activating protein; ADP-ribosylation factor-directed GTPase-activating protein 1; AMAP1; ARF GTPase-activating protein 1; arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain 1; ArfGAP with SH3 domain, ankyrin repeat and PH domain1; Asap1; AV239055; centaurin, beta 4; CENTB4; Ddef1; DEF-1; development and differentiation enhancing factor 1; development and differentiation-enhancing factor 1; differentiation enhancing factor 1; differentiation-enhancing factor 1; Kiaa1249; mKIAA1249; PAG2; PAP; PIP2-dependent ARF1 GAP; s19; Shag1; src SH3 binding protein; ZG14P | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULH1 | |
| ASAP1 | |
| Recombinant protein corresponding to Human ASAP1. Recombinant protein control fragment (Product #RP-104108). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction