missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ASAP1 Recombinant Protein Antigen

Product Code. 18667177 Shop All Bio Techne Products
Click to view available options
:
0.1 mL
This item is not returnable. View return policy

Product Code. 18667177

Brand: Novus Biologicals™ NBP248519PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1 M Urea, pH 7.4.
Gene ID (Entrez) 50807
Molecular Weight (g/mol) MolecularWeight-theroretical: 27 kDa
Quantity 0.1 mL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Gene Alias 130 kDa phosphatidylinositol 4,5-biphosphate-dependent ARF1 GTPase-activatingprotein, AMAP1, ARF GTPase-activating protein 1, arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 1, ArfGAP with SH3 domain, ankyrin repeat and PH domain 1, centaurin, beta 4, CENTB4, DDEF1ADP-ribosylation factor-directed GTPase-activating protein 1, development and differentiation enhancing factor 1, Development and differentiation-enhancing factor 1, Differentiation-enhancing factor 1, KIAA1249DEF-1, PAG2, PAP, PIP2-dependent ARF1 GAP, ZG14P
Gene Symbol ASAP1
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-48519. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mLFor current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking peptide related information and a protocol, click here .
Research Category Phospho Specific
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein ASAP1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.