missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASAP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17107353
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17107353 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17107353 Supplier Invitrogen™ Supplier No. PA5109417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MPDQISVSEF VAETHEDYKA PTASSFTTRT AQCRNTVAAI EEALDVDRMV LYKMKKSVKA INSSGLAHVE NEEQYTQALE KFGGNCVCRD DPDLGSAFLK FSVFTKELTA LFKNLIQNMN.

ASAP2 is a multidomain protein belonging to the beta family with two ANK repeats, an Arf-GAP domain, a PH domain and a SH3 domain. This protein localizes in the Golgi apparatus and also at the plasma membrane where it colocalizes with protein tyrosine kinase 2-beta (PYK2) and forms a stable complex with PYK2 in vivo. ASAP2 is tyrosine phosphorylated by activated PYK2. It functions as a substrate and downstream target for PYK2 and SRC, a pathway that may be involved in the regulation of vesicular transport. ASAP2 activates the small GTPases ARF1, ARF5 and ARF6 and regulates the formation of post-Golgi vesicles and modulates constitutive secretion. It modulates phagocytosis mediated by Fc gamma receptor and ARF6. Usually detected in heart, brain, placenta, kidney, monocytes and pancreas, it is up-regulated during monocyte maturation. Multiple transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASAP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ASAP2
Gene Accession No. Q7SIG6
Gene Alias 6530401G17Rik; AMAP2; Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2; ArfGAP with SH3 domain, ankyrin repeat and PH domain 2; Asap2; centaurin, beta 3; CENTB3; DDEF2; development and differentiation enhancing factor 2; development and differentiation-enhancing factor 2; Gm1523; Gm592; KIAA0400; PAG3; PAP; Pap-alpha; PAPbeta; Paxillin-associated protein with ARF GAP activity 3; PYK2 C terminus-associated protein; pyk2 C-terminus-associated protein; SHAG1
Gene Symbols ASAP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human ASAP2 (NP_001128663).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 211914
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.