missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15889063
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15889063 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15889063 Supplier Invitrogen™ Supplier No. PA570658

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 93%.

The protein encoded by this gene contains an ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signaling (SOCS) proteins and their binding partners with the elongin B and C complex, targeting them for ubiquitination and degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ASB1
Gene Accession No. Q9Y576
Gene Alias 1700029O08Rik; 1700054C17Rik; AI426616; AI851378; ankyrin repeat and SOCS box containing 1; ankyrin repeat and SOCS box protein 1; ankyrin repeat and SOCS box-containing 1; ankyrin repeat and SOCS box-containing protein 1; ankyrin repeat-containing SOCS box protein 1; ASB1; ASB-1; KIAA1146; LOW QUALITY PROTEIN: ankyrin repeat and SOCS box protein 1; mKIAA1146
Gene Symbols ASB1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ASB1 (aa 236-285).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51665
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.