missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB13 Monoclonal Antibody (OTI7E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809435S
This item is not returnable.
View return policy
Description
ASB13 Monoclonal antibody specifically detects ASB13 in Human samples. It is validated for Western BlotSpecifications
| ASB13 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WXK3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79754 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7E10 | |
| Unconjugated | |
| ASB13 | |
| 2210015B19Rik; 6430573K02Rik; ankyrin repeat and SOCS box containing 13; ankyrin repeat and SOCS box protein 13; ankyrin repeat and SOCS box-containing 13; ankyrin repeat and SOCS box-containing protein 13; ankyrin repeat domain-containing SOCS box protein 13; ankyrin repeat domain-containing SOCS box protein Asb-13; ASB13; Asb-13; C85285 | |
| Human recombinant protein fragment corresponding to amino acids 76-278 of human ASB13 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction