missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB5 Antibody (6B10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00140458-M03-100ug
This item is not returnable.
View return policy
Description
ASB5 Monoclonal antibody specifically detects ASB5 in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| ASB5 | |
| Monoclonal | |
| Unconjugated | |
| NP_543150 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA | |
| 6B10 | |
| In 1x PBS, pH 7.4 | |
| ankyrin repeat and SOCS box containing 5, ankyrin repeat and SOCS box protein 5, ankyrin repeat and SOCS box-containing 5, ASB-5, SOCS box protein ASB-5 | |
| ASB5 (NP_543150, 220 a.a. ∽ 328 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LLYAGADVQKGKYWDTPLHAAAQQSSTEIVNLLLEFGADINAKNTELLRPIDVATSSSMVERILLQHEATPSSLYQLCRLCIRSYIGKPRLHLIPQLQLPTLLKNFLQY | |
| 100 μg | |
| Signal Transduction | |
| 140458 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction