missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ASB6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15740031 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15740031 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15740031 Supplier Invitrogen™ Supplier No. PA552077

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: IIFEYQPLVD AILGSLGIQD PERQESLDRP SYVASEESRI LVLTELLERK AHSPFYQEGV SNALLKMAEL GLTRAADVLL RHGANLNFED PVTYYTALHI AVLRNQPDMV ELLVHHGADV NRRD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 43%.

ASB6 belongs to a family of ankyrin repeat proteins that, along with four other protein families, contain a C-terminal SOCS box motif. Growing evidence suggests that the SOCS box, similar to the F-box, acts as a bridge between specific substrate-binding domains and the more generic proteins that comprise a large family of E3 ubiquitin protein ligases.The protein encoded by this gene belongs to a family of ankyrin repeat proteins that, along with four other protein families, contain a C-terminal SOCS box motif. Growing evidence suggests that the SOCS box, similar to the F-box, acts as a bridge between specific substrate-binding domains and the more generic proteins that comprise a large family of E3 ubiquitin protein ligases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASB6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ASB6
Gene Accession No. Q9NWX5
Gene Alias ankyrin repeat and SOCS box containing 6; ankyrin repeat and SOCS box protein 6; ankyrin repeat and SOCS box-containing 6; ASB6; ASB-6
Gene Symbols ASB6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ASB6. Recombinant protein control fragment (Product #RP-102084).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140459
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.