missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASB9 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98662JF549
This item is not returnable.
View return policy
Description
ASB9 Polyclonal specifically detects ASB9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ASB9 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 549 | |
| ankyrin repeat and SOCS box containing 9, ankyrin repeat and SOCS box protein 9, ankyrin repeat and SOCS box-containing 9, ankyrin repeat and suppressor of cytokine signaling box protein 9, ASB-9, DKFZP564L0862, FLJ20636, MGC4954 | |
| Produced in rabbits immunized with E. coli-derived Human ASB9 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 140462 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction