missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASC/TMS1 Antibody (OTI3E9), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71853AF350
This item is not returnable.
View return policy
Description
ASC/TMS1 Monoclonal specifically detects ASC/TMS1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| ASC/TMS1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PYCARD | |
| This ASC/TMS1 Antibody (OTI3E9) was developed against full length human recombinant protein of human ASC/TMS1 (NP_037390) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI3E9 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| ASCMGC10332, CARD5TMS, caspase recruitment domain protein 5, Caspase recruitment domain-containing protein 5, hASC, PYD and CARD domain containing, PYD and CARD domain-containing protein, Target of methylation-induced silencing 1, TMS-1, TMS1apoptosis-associated speck-like protein containing a CARD | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer, Cell Biology, Inflammation, Innate Immunity | |
| 29108.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction