missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ASC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30189
This item is not returnable.
View return policy
Description
ASC1 Polyclonal antibody specifically detects ASC1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ASC1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| activating signal cointegrator 1, ASC-1, HsT17391, thyroid hormone receptor interactor 4, thyroid receptor interacting protein 4, Thyroid receptor-interacting protein 4, TR-interacting protein 4, TRIP-4 | |
| between 225 and 275 | |
| 100 μg | |
| Apoptosis, Cancer, DNA Repair, Tumor Suppressors | |
| 9325 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction