missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCC1 Antibody (OTI6G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01345
This item is not returnable.
View return policy
Description
ASCC1 Monoclonal specifically detects ASCC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ASCC1 | |
| Monoclonal | |
| 0.41 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| ASCC1 | |
| Full length human recombinant protein of human ASCC1(NP_057031) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 51008 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI6G5 | |
| Unconjugated | |
| activating signal cointegrator 1 complex subunit 1, ASC-1 complex subunit p50, ASC1p50, CGI-18, Em:AC022392.3, RP11-150D20.4, Trip4 complex subunit p50 | |
| Mouse | |
| 41 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction