missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASCC1 Antibody (OTI6G5), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71879PECY55
This item is not returnable.
View return policy
Description
ASCC1 Monoclonal specifically detects ASCC1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| ASCC1 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| ASCC1 | |
| Full length recombinant protein of human ASCC1 (NP_057031) produced in HEK293T cells. | |
| 0.1 mL | |
| 51008.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI6G5 | |
| Flow Cytometry | |
| activating signal cointegrator 1 complex subunit 1, ASC-1 complex subunit p50, ASC1p50, CGI-18, Em:AC022392.3, RP11-150D20.4, Trip4 complex subunit p50 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction